Learn More
Abnova™ Human TSFM Partial ORF (NP_005717, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010102-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes. The TSFM gene encodes a mitochondrial translation elongation factor (Smeitink et al., 2006 [PubMed 17033963]).[supplied by OMIM]
Sequence: MSLLRSLRVFLVARTGSYPAGSLLRQSPQPRHTFYAGPRLSASASSKELLMKLRRKTGYSFVNCKKALETCGGDLKQAEIWLHKEAQKEGWSKAAKLQGRSpecifications
NP_005717 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSLLRSLRVFLVARTGSYPAGSLLRQSPQPRHTFYAGPRLSASASSKELLMKLRRKTGYSFVNCKKALETCGGDLKQAEIWLHKEAQKEGWSKAAKLQGR | |
RUO | |
TSFM | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10102 | |
TSFM (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COXPD3/EF-TS/EF-Tsmt | |
TSFM | |
Recombinant | |
wheat germ expression system |