missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRUB2 Full-length ORF (NP_056494.1, 1 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026995-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16158452

  • 335.00€ / 10µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

Pseudouridine is an abundant component of rRNAs and tRNAs and is enzymatically generated by isomerization of uridine by pseudouridine synthase (Zucchini et al., 2003 [PubMed 12736709]).[supplied by OMIM]

Sequence: MGSAGLSRLHGLFAVYKPPGLKWKHLRDTVELQLLKGLNARKPPAPKQRVRFLLGPMEGSEEKELTLTATSVPSFINHPLVCGPAFAHLKVGVGHRLDAQASGVLVLGVGHGCRLLTDMYNAHLTKDYTVRGLLGKATDDFREDGRLVEKTTYDHVTREKLDRILAVIQGSHQKALVMYSNLDLKTQEAYEMAVRGLIRPMNKSPMLITGIRCLYFAPPEFLLEVQCMHETQKELRKLVHEIGLELKTTAVCTQVRRTRDGFFTLDSALLRTQWDLTNIQDAIRAATPQVAAELEKSLSPGLDTKQLPSPGWSWDSQGPSSTLGLERGAGQ
Specifications

Specifications

NP_056494.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
63.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CLONE24922/RP11-339B21.1
TRUB2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
26995
TRUB2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSAGLSRLHGLFAVYKPPGLKWKHLRDTVELQLLKGLNARKPPAPKQRVRFLLGPMEGSEEKELTLTATSVPSFINHPLVCGPAFAHLKVGVGHRLDAQASGVLVLGVGHGCRLLTDMYNAHLTKDYTVRGLLGKATDDFREDGRLVEKTTYDHVTREKLDRILAVIQGSHQKALVMYSNLDLKTQEAYEMAVRGLIRPMNKSPMLITGIRCLYFAPPEFLLEVQCMHETQKELRKLVHEIGLELKTTAVCTQVRRTRDGFFTLDSALLRTQWDLTNIQDAIRAATPQVAAELEKSLSPGLDTKQLPSPGWSWDSQGPSSTLGLERGAGQ
RUO
TRUB2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers