missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRIP Partial ORF (NP_005870, 370 a.a. - 469 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010293-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16164842

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein that contains an N-terminal RING finger motif and a putative coiled-coil domain. A similar murine protein interacts with TNFR-associated factor 1 (TRAF1), TNFR-associated factor 2 (TRAF2), and cylindromatosis. The interaction with TRAF2 inhibits TRAF2-mediated nuclear factor kappa-B, subunit 1 activation that is required for cell activation and protection against apoptosis. [provided by RefSeq]

Sequence: GQSCAGEPDEELVGAFPIFVRNAILGQKQPKRPRSESSCSKDVVRTGFDGLGGRTKFIQPTDTVMIRPLPVKPKTKVKQRVRVKTVPSLFQAKLDTFLWS
Specifications

Specifications

NP_005870
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GQSCAGEPDEELVGAFPIFVRNAILGQKQPKRPRSESSCSKDVVRTGFDGLGGRTKFIQPTDTVMIRPLPVKPKTKVKQRVRVKTVPSLFQAKLDTFLWS
RUO
TRAIP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10293
TRIP (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RNF206/TRIP
TRAIP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers