missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRAP1 Full-length ORF (NP_057376.1, 1 a.a. - 704 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010131-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194532

  • 349.00€ / 10µg

Estimated Shipment: 03-11-2023
to see stock.
Add to basket

Description

Description

HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. TRAP1 is a mitochondrial HSP90 protein. Other HSP90 proteins are found in cytosol (see HSP90AA1; MIM 140571) and endoplasmic reticulum (HSP90B1; MIM 191175) (Chen et al., 2005 [PubMed 16269234]).[supplied by OMIM]

Sequence: MARELRALLLWGRRLRPLLRAPALAAVPGGKPILCPRRTTAQLGPRRNPAWSLQAGRLFSTQTAEDKEEPLHSIISSTESVQGSTSKHEFQAETKKLLDIVARSLYSEKEVFIRELISNASDALEKLRHKLVSDGQALPEMEIHLQTNAEKGTITIQDTGIGMTQEELVSNLGTIARSGSKAFLDALQNQAEASSKIIGQFGVGFYSAFMVADRVEVYSRSAAPGSLGYQWLSDGSGVFEIAEASGVRTGTKIIIHLKSDCKEFSSEARVRDVVTKYSNFVSFPLYLNGRRMNTLQAIWMMDPKDVGEWQHEEFYRYVAQAHDKPRYTLHYKTDAPLNIRSIFYVPDMKPSMFDVSRELGSSVALYSRKVLIQTKATDILPKWLRFIRGVVDSEDIPLNLSRELLQESALIRKLRDVLQQRLIKFFIDQSKKDAEKYAKFFEDYGLFMREGIVTATEQEVKEDIAKLLRYESSALPSGQLTSLSEYASRMRAGTRNIYYLCAPNRHLAEHSPYYEAMKKKDTEVLFCFEQFDELTLLHLREFDKKKLISVETDIVVDHYKEEKFEDRSPAAECLSEKETEELMAWMRNVLGSRVTNVKVTLRLDTHPAMVTVLEMGAARHFLRMQQLAKTQEERAQLLQPTLEINPRHALIKKLNQLRASEPGLAQLLVDQIYENAMIAAGLVDDPRAMVGRLNELLVKALERH
Specifications

Specifications

NP_057376.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
106.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSP75/HSP90L
TRAP1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10131
TRAP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MARELRALLLWGRRLRPLLRAPALAAVPGGKPILCPRRTTAQLGPRRNPAWSLQAGRLFSTQTAEDKEEPLHSIISSTESVQGSTSKHEFQAETKKLLDIVARSLYSEKEVFIRELISNASDALEKLRHKLVSDGQALPEMEIHLQTNAEKGTITIQDTGIGMTQEELVSNLGTIARSGSKAFLDALQNQAEASSKIIGQFGVGFYSAFMVADRVEVYSRSAAPGSLGYQWLSDGSGVFEIAEASGVRTGTKIIIHLKSDCKEFSSEARVRDVVTKYSNFVSFPLYLNGRRMNTLQAIWMMDPKDVGEWQHEEFYRYVAQAHDKPRYTLHYKTDAPLNIRSIFYVPDMKPSMFDVSRELGSSVALYSRKVLIQTKATDILPKWLRFIRGVVDSEDIPLNLSRELLQESALIRKLRDVLQQRLIKFFIDQSKKDAEKYAKFFEDYGLFMREGIVTATEQEVKEDIAKLLRYESSALPSGQLTSLSEYASRMRAGTRNIYYLCAPNRHLAEHSPYYEAMKKKDTEVLFCFEQFDELTLLHLREFDKKKLISVETDIVVDHYKEEKFEDRSPAAECLSEKETEELMAWMRNVLGSRVTNVKVTLRLDTHPAMVTVLEMGAARHFLRMQQLAKTQEERAQLLQPTLEINPRHALIKKLNQLRASEPGLAQLLVDQIYENAMIAAGLVDDPRAMVGRLNELLVKALERH
RUO
TRAP1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers