Learn More
R&D Systems™ Human Transglutaminase 3/TGM3 (NP_003236.3) VersaClone cDNA
Product Code. 18754267
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18754267 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18754267 Supplier R&D Systems™ Supplier No. RDC3200
  • ONLINE EXCLUSIVE PRICE
    761.00€
    719.25€ / 10µg
    Save 41.75€
    5% Off
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
Contains gene inserts that are fully sequenced and produced with high quality
TRUSTED_SUSTAINABILITY
Specifications
Gene Transglutaminase 3/TGM3
Accession Number NP_003236.3
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 2095 bp
Gene ID (Entrez) 7053
Gene Alias EC 2.3.2.13, MGC126250, protein-glutamine gamma-glutamyltransferase E, TG(E), TG3, TGase E, TGase-3, TgaseE, TGEMGC126249, TGM3, transglutaminase 3 (E polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase E, transglutaminase-3
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg