Learn More
Abcam Human TRANK1 knockout A549 cell line
Product Code. 32140902 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32140902 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32140902 Supplier Abcam Supplier No. ab2671872x1000000Cells_
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
TRANK1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp insertion in exon 11. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout cells: selec...
TRANK1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp insertion in exon 11. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells