missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRAF7 Partial ORF (AAH24267, 61 a.a. - 160 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084231-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100697

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

Tumor necrosis factor (TNF; see MIM 191160) receptor-associated factors, such as TRAF7, are signal transducers for members of the TNF receptor superfamily (see MIM 191190). TRAFs are composed of an N-terminal cysteine/histidine-rich region containing zinc RING and/or zinc finger motifs; a coiled-coil (leucine zipper) motif; and a homologous region that defines the TRAF family, the TRAF domain, which is involved in self-association and receptor binding.[supplied by OMIM]

Sequence: VFKDPVITTCGHTFCRRCALKSEKCPVDNVKLTVVVNNIAVAEQIGELFIHCRHGCRVAGSGKPPIFEVDPRGCPFTIKLSARKDHEGSCDYRPVRCPNN
Specifications

Specifications

AAH24267
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VFKDPVITTCGHTFCRRCALKSEKCPVDNVKLTVVVNNIAVAEQIGELFIHCRHGCRVAGSGKPPIFEVDPRGCPFTIKLSARKDHEGSCDYRPVRCPNN
RUO
TRAF7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
84231
TRAF7 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp586I021/MGC7807/RFWD1/RNF119
TRAF7
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers