missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRAF5 Full-length ORF (NP_001029082.1, 1 a.a. - 557 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007188-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193798

  • 528.00€ / 25µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

The scaffold protein encoded by this gene is a member of the tumor necrosis factor receptor-associated factor (TRAF) protein family and contains a meprin and TRAF homology (MATH) domain, a RING-type zinc finger, and two TRAF-type zinc fingers. TRAF proteins are associated with, and mediate signal transduction from members of the TNF receptor superfamily. This protein is one of the components of a multiple protein complex which binds to tumor necrosis factor (TNF) receptor cytoplasmic domains and mediates TNF-induced activation. Alternate transcriptional splice variants have been characterized. [provided by RefSeq]

Sequence: MAYSEEHKGMPCGFIRQNSGNSISLDFEPSIEYQFVERLEERYKCAFCHSVLHNPHQTGCGHRFCQHCILSLRELNTVPICPVDKEVIKSQEVFKDNCCKREVLNLYVYCSNAPGCNAKVILGRYQDHLQQCLFQPVQCSNEKCREPVLRKDLKEHLSASCQFRKEKCLYCKKDVVVINLQNHEENLCPEYPVFCPNNCAKIILKTEVDEHLAVCPEAEQDCPFKHYGCAVTDKRRNLQQHEHSALREHMRLVLEKNVQLEEQISDLHKSLEQKESKIQQLAETIKKLEKEFKQFAQLFGKNGSFLPNIQVFASHIDKSAWLEAQVHQLLQMVNQQQNKFDLRPLMEAVDTVKQKITLLENNDQRLAVLEEETNKHDTHINIHKAQLSKNEERFKLLEGTCYNGKLIWKVTDYKMKKREAVDGHTVSIFSQSFYTSRCGYRLCARAYLNGDGSGRGSHLSLYFVVMRGEFDSLLQWPFRQRVTLMLLDQSGKKNIMETFKPDPNSSSFKRPDGEMNIASGCPRFVAHSVLENAKNAYIKDDTLFLKVAVDLTDLEDL
Specifications

Specifications

NP_001029082.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
90.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC:39780/RNF84
TRAF5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7188
TRAF5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAYSEEHKGMPCGFIRQNSGNSISLDFEPSIEYQFVERLEERYKCAFCHSVLHNPHQTGCGHRFCQHCILSLRELNTVPICPVDKEVIKSQEVFKDNCCKREVLNLYVYCSNAPGCNAKVILGRYQDHLQQCLFQPVQCSNEKCREPVLRKDLKEHLSASCQFRKEKCLYCKKDVVVINLQNHEENLCPEYPVFCPNNCAKIILKTEVDEHLAVCPEAEQDCPFKHYGCAVTDKRRNLQQHEHSALREHMRLVLEKNVQLEEQISDLHKSLEQKESKIQQLAETIKKLEKEFKQFAQLFGKNGSFLPNIQVFASHIDKSAWLEAQVHQLLQMVNQQQNKFDLRPLMEAVDTVKQKITLLENNDQRLAVLEEETNKHDTHINIHKAQLSKNEERFKLLEGTCYNGKLIWKVTDYKMKKREAVDGHTVSIFSQSFYTSRCGYRLCARAYLNGDGSGRGSHLSLYFVVMRGEFDSLLQWPFRQRVTLMLLDQSGKKNIMETFKPDPNSSSFKRPDGEMNIASGCPRFVAHSVLENAKNAYIKDDTLFLKVAVDLTDLEDL
RUO
TRAF5
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers