missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total MMP-1 DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 15 Plates)
Unit Size:
Pack of 15
Description
The Human Total MMP-1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human MMP-1.
Specifications
Specifications
| Assay Range | 62.50 to 4000 pg/mL |
| Immunoassay Kit Format | 96-well Plate |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
| Sufficient For | Fifteen 96-well plates |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 4312 |
| Kit Contents | 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer |
| Quantity | 1 Kit (for 15 Plates) |
| Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction