Learn More
Human TOB1 (TOB) knockout HeLa cell lysate, 1 Kit
Product Code. 32141060 Shop All Abcam Products
Product Code. 32141060 Supplier Abcam Supplier No. ab2587291Kit
 Restricted Item
Estimated Shipment: 09-12-2026
Human TOB1 (TOB) knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: TOB1 (TOB) knockout HeLa