missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TNFRSF9 Partial ORF (NP_001552.2, 156 a.a. - 255 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003604-Q04.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16094265

  • 508.00€ / 25µg
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor contributes to the clonal expansion, survival, and development of T cells. It can also induce proliferation in peripheral monocytes, enhance T cell apoptosis induced by TCR/CD3 triggered activation, and regulate CD28 co-stimulation to promote Th1 cell responses. The expression of this receptor is induced by lymphocyte activation. TRAF adaptor proteins have been shown to bind to this receptor and transduce the signals leading to activation of NF-kappaB. [provided by RefSeq]

Sequence: VVCGPSPADLSPGASSVTPPAPAREPGHSPQIISFFLALTSTALLFLLFFLTLRFSVVKRGRKKLLYIFKQPFMRPVQTTQEEDGCSCRFPEEEEGGCEL
Specifications

Specifications

NP_001552.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue
VVCGPSPADLSPGASSVTPPAPAREPGHSPQIISFFLALTSTALLFLLFFLTLRFSVVKRGRKKLLYIFKQPFMRPVQTTQEEDGCSCRFPEEEEGGCEL
RUO
TNFRSF9
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3604
TNFRSF9 (Human) Recombinant Protein (Q04)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
4-1BB/CD137/CDw137/ILA/MGC2172
TNFRSF9
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers