missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TNFRSF13C Full-length ORF (NP_443177.1, 1 a.a. - 184 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00115650-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157233

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

B cell-activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. Overexpression of Baff in mice results in mature B-cell hyperplasia and symptoms of systemic lupus erythematosus (SLE). Also, some SLE patients have increased levels of BAFF in serum. Therefore, it has been proposed that abnormally high levels of BAFF may contribute to the pathogenesis of autoimmune diseases by enhancing the survival of autoreactive B cells. The protein encoded by this gene is a receptor for BAFF and is a type III transmembrane protein containing a single extracellular cysteine-rich domain. It is thought that this receptor is the principal receptor required for BAFF-mediated mature B-cell survival. [provided by RefSeq]

Sequence: MRRGPRSLRGRDAPAPTPCVPAECFDLLVRHCVACGLLRTPRPKPAGASSPAPRTALQPQESVGAGAGEAALPLPGLLFGAPALLGLALVLALVLVGLVSWRRRQRRLRGASSAEAPDGDKDAPEPLDKVIILSPGISDATAPAWPPPGEDPGTTPPGHSVPVPATELGSTELVTTKTAGPEQQ
Specifications

Specifications

NP_443177.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BAFF-R/BAFFR/CD268/MGC138235
TNFRSF13C
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
115650
TNFRSF13C (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRRGPRSLRGRDAPAPTPCVPAECFDLLVRHCVACGLLRTPRPKPAGASSPAPRTALQPQESVGAGAGEAALPLPGLLFGAPALLGLALVLALVLVGLVSWRRRQRRLRGASSAEAPDGDKDAPEPLDKVIILSPGISDATAPAWPPPGEDPGTTPPGHSVPVPATELGSTELVTTKTAGPEQQ
RUO
TNFRSF13C
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers