Learn More
Abnova™ Human TNFAIP1 Partial ORF (NP_066960, 258 a.a. - 316 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007126-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. Studies of a similar gene in mouse suggest that the expression of this gene is developmentally regulated in a tissue-specific manner. [provided by RefSeq]
Sequence: TPRVPDNSLLEATSRSRSQASPSEDEETFELRDRVRRIHVKRYSTYDDRQLGHQSTHRDSpecifications
NP_066960 | |
Liquid | |
7126 | |
TNFAIP1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B12/B61/EDP1/MGC2317 | |
TNFAIP1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.23kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TPRVPDNSLLEATSRSRSQASPSEDEETFELRDRVRRIHVKRYSTYDDRQLGHQSTHRD | |
RUO | |
TNFAIP1 | |
Wheat Germ (in vitro) | |
GST |