missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TNF RII/TNFRSF1B Magnetic Luminex Performance Assay

Product Code. 30001965
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001965 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001965 Supplier R&D Systems™ Supplier No. LUCAM726

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Simultaneous quantitative determination of multiple analytes.

  • Magnetic format allows for easier wash steps
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
  • Requires a small sample volume (<50μL)
  • Run your assay in just 3-5 hours
  • Mass-calibrated standards for consistent results with every new lot of material
  • Enough reagents for one 96-well plate
TRUSTED_SUSTAINABILITY

Specifications

Product Type Magnetic Bead-based Assay
Quantity 100 Tests
Species Human
Detection Method Fluorescence
Format 96-well Microplate and Antibody-coated Magnetic Beads
For Use With (Application) Analysis of biochemical parameters
For Use With (Equipment) Luminex MAGPIX Platform
Kit Contents Bead Diluents, Biotin Antibody Diluents, Calibrator Diluents, Foil Plate Sealers, Mixing Bottles, Round-bottom Microplate, Standard Cocktail(s), Streptavidin-PE, Wash Buffer Concentrate
Content And Storage Store the unopened product at 2° to 8°C. Do not use past expiration date.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.