missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TNF RI/TNFRSF1A Quantikine ELISA Kit

Product Code. p-4753699
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Sufficient For unitSize
18752024 1 Plate 1 x 96-well Plate 1 set
18762104 6 Plates 6 x 96-well Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18752024 Supplier R&D Systems™ Supplier No. DRT100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human TNF RI/TNFRSF1A Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human TNF RI/TNFRSF1A Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human TNF RI/TNFRSF1A in cell culture supernates (200 ul), serum (20 ul), edta plasma (20 ul), heparin plasma (20 ul), citrate plasma (20 ul), urine (20 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 7.80 - 500pg/mL
Assay Sensitivity 1.2pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine
Specificity Natural and Recombinant Human sTNF RI
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 7132
Quantity 1 Plate
Sample Volume Cell Culture Supernates (200uL),Serum (20uL),EDTA Plasma (20uL),Heparin Plasma (20uL),Citrate Plasma (20uL),Urine (20uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNFRI, TNF-RI, TNF-R-I, TNFR-I, TNFRSF1A, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1 A isoform beta, tumor necrosis factor receptor superfamily member 1 A, tumor necrosis factor receptor superfamily, member 1 A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.