Learn More
Abnova™ Human TMPRSS5 Full-length ORF (NP_110397.1, 1 a.a. - 457 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00080975-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that belongs to the serine protease family. Serine proteases are known to be involved in many physiological and pathological processes. [provided by RefSeq]
Sequence: MSLMLDDQPPMEAQYAEEGPGPGIFRAEPGDQQHPISQAVCWRSMRRGCAVLGALGLLAGAGVGSWLLVLYLCPAASQPISGTLQDEEITLSCSEASAEEALLPALPKTVSFRINSEDFLLEAQVRDQPRWLLVCHEGWSPALGLQICWSLGHLRLTHHKGVNLTDIKLNSSQEFAQLSPRLGGFLEEAWQPRNNCTSGQVVSLRCSECGARPLASRIVGGQSVAPGRWPWQASVALGFRHTCGGSVLAPRWVVTAAHCMHSFRLARLSSWRVHAGLVSHSAVRPHQGALVERIIPHPLYSAQNHDYDVALLRLQTALNFSDTVGAVCLPAKEQHFPKGSRCWVSGWGHTHPSHTYSSDMLQDTVVPLFSTQLCNSSCVYSGALTPRMLCAGYLDGRADACQGDSGGPLVCPDGDTWRLVGVVSWGRGCAEPNHPGVYAKVAEFLDWIHDTAQDSLLSpecifications
NP_110397.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC141886/MGC148044/SPINESIN | |
TMPRSS5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
80975 | |
TMPRSS5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSLMLDDQPPMEAQYAEEGPGPGIFRAEPGDQQHPISQAVCWRSMRRGCAVLGALGLLAGAGVGSWLLVLYLCPAASQPISGTLQDEEITLSCSEASAEEALLPALPKTVSFRINSEDFLLEAQVRDQPRWLLVCHEGWSPALGLQICWSLGHLRLTHHKGVNLTDIKLNSSQEFAQLSPRLGGFLEEAWQPRNNCTSGQVVSLRCSECGARPLASRIVGGQSVAPGRWPWQASVALGFRHTCGGSVLAPRWVVTAAHCMHSFRLARLSSWRVHAGLVSHSAVRPHQGALVERIIPHPLYSAQNHDYDVALLRLQTALNFSDTVGAVCLPAKEQHFPKGSRCWVSGWGHTHPSHTYSSDMLQDTVVPLFSTQLCNSSCVYSGALTPRMLCAGYLDGRADACQGDSGGPLVCPDGDTWRLVGVVSWGRGCAEPNHPGVYAKVAEFLDWIHDTAQDSLL | |
RUO | |
TMPRSS5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |