Learn More
Abnova™ Human TMPRSS11D Full-length ORF (NP_004253.1, 1 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009407-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a trypsin-like serine protease released from the submucosal serous glands onto mucous membrane. It is a type II integral membrane protein and has 29-38% identity in the sequence of the catalytic region with human hepsin, enteropeptidase, acrosin, and mast cell tryptase. The noncatalytic region has little similarity to other known proteins. This protein may play some biological role in the host defense system on the mucous membrane independently of or in cooperation with other substances in airway mucous or bronchial secretions. [provided by RefSeq]
Sequence: MYRPARVTSTSRFLNPYVVCFIVVAGVVILAVTIALLVYFLAFDQKSYFYRSSFQLLNVEYNSQLNSPATQEYRTLSGRIESLITKTFKESNLRNQFIRAHVAKLRQDGSGVRADVVMKFQFTRNNNGASMKSRIESVLRQMLNNSGNLEINPSTEITSLTDQAAANWLINECGAGPDLITLSEQRILGGTEAEEGSWPWQVSLRLNNAHHCGGSLINNMWILTAAHCFRSNSNPRDWIATSGISTTFPKLRMRVRNILIHNNYKSATHENDIALVRLENSVTFTKDIHSVCLPAATQNIPPGSTAYVTGWGAQEYAGHTVPELRQGQVRIISNDVCNAPHSYNGAILSGMLCAGVPQGGVDACQGDSGGPLVQEDSRRLWFIVGIVSWGDQCGLPDKPGVYTRVTAYLDWIRQQTGISpecifications
NP_004253.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HAT/MGC150587/MGC150588 | |
TMPRSS11D | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9407 | |
TMPRSS11D (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MYRPARVTSTSRFLNPYVVCFIVVAGVVILAVTIALLVYFLAFDQKSYFYRSSFQLLNVEYNSQLNSPATQEYRTLSGRIESLITKTFKESNLRNQFIRAHVAKLRQDGSGVRADVVMKFQFTRNNNGASMKSRIESVLRQMLNNSGNLEINPSTEITSLTDQAAANWLINECGAGPDLITLSEQRILGGTEAEEGSWPWQVSLRLNNAHHCGGSLINNMWILTAAHCFRSNSNPRDWIATSGISTTFPKLRMRVRNILIHNNYKSATHENDIALVRLENSVTFTKDIHSVCLPAATQNIPPGSTAYVTGWGAQEYAGHTVPELRQGQVRIISNDVCNAPHSYNGAILSGMLCAGVPQGGVDACQGDSGGPLVQEDSRRLWFIVGIVSWGDQCGLPDKPGVYTRVTAYLDWIRQQTGI | |
RUO | |
TMPRSS11D | |
Wheat Germ (in vitro) | |
GST | |
Liquid |