Learn More
R&D Systems™ Human TMPRSS11A (BAD18660.1) VersaClone cDNA
Specifications
Specifications
| Gene | TMPRSS11A |
| Accession Number | BAD18660.1 |
| Gene ID (Entrez) | 339967 |
| Gene Alias | Airway Trypsin-Like Protease 1, EC 3.4.21, EC 3.4.21., ECRG1, Epidermal Type II Transmembrane Serine Protease, Epidermal Type-II Transmembrane Serine Protease, Esophageal Cancer-Susceptibility Gene 1 Protein, HATL1, HESP, Transmembrane Protease Serine 11 A, Transmembrane Protease, Serine 11 A |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |