missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TMEM167B Full-length ORF (NP_064526.1, 1 a.a. - 74 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00056900-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122313

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MTNVYSLDGILVFGLLFVCTCAYFKKVPRLKTWLLSEKKGVWGVFYKAAVIGTRLHAAVAIACVVMAFYVLFIK
Specifications

Specifications

NP_064526.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.54kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AD-020/C1orf119/FLJ90710
TMEM167B
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
56900
TMEM167B (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue
MTNVYSLDGILVFGLLFVCTCAYFKKVPRLKTWLLSEKKGVWGVFYKAAVIGTRLHAAVAIACVVMAFYVLFIK
RUO
TMEM167B
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers