Description
TMEM106B KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9 X = 1 bp insertion Frameshift: 100%. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Quantity | 2 x 1,000,000 Cells |
| Regulatory Status | RUO |