missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TIMP4 Full-length ORF (NP_003247.1, 1 a.a. - 224 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007079-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123658

  • 335.00€ / 10µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

This gene belongs to the TIMP gene family. The proteins encoded by this gene family are inhibitors of the matrix metalloproteinases, a group of peptidases involved in degradation of the extracellular matrix. The secreted, netrin domain-containing protein encoded by this gene is involved in regulation of platelet aggregation and recruitment and may play role in hormonal regulation and endometrial tissue remodeling. [provided by RefSeq]

Sequence: MPGSPRPAPSWVLLLRLLALLRPPGLGEACSCAPAHPQQHICHSALVIRAKISSEKVVPASADPADTEKMLRYEIKQIKMFKGFEKVKDVQYIYTPFDSSLCGVKLEANSQKQYLLTGQVLSDGKVFIHLCNYIEPWEDLSLVQRESLNHHYHLNCGCQITTCYTVPCTISAPNECLWTDWLLERKLYGYQAQHYVCMKHVDGTCSWYRGHLPLRKEFVDIVQP
Specifications

Specifications

NP_003247.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TIMP4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
7079
TIMP4 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPGSPRPAPSWVLLLRLLALLRPPGLGEACSCAPAHPQQHICHSALVIRAKISSEKVVPASADPADTEKMLRYEIKQIKMFKGFEKVKDVQYIYTPFDSSLCGVKLEANSQKQYLLTGQVLSDGKVFIHLCNYIEPWEDLSLVQRESLNHHYHLNCGCQITTCYTVPCTISAPNECLWTDWLLERKLYGYQAQHYVCMKHVDGTCSWYRGHLPLRKEFVDIVQP
RUO
TIMP4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers