Learn More
Abnova™ Human TIMP4 Full-length ORF (AAH10553.1, 1 a.a. - 224 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007079-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the TIMP gene family. The proteins encoded by this gene family are inhibitors of the matrix metalloproteinases, a group of peptidases involved in degradation of the extracellular matrix. The secreted, netrin domain-containing protein encoded by this gene is involved in regulation of platelet aggregation and recruitment and may play role in hormonal regulation and endometrial tissue remodeling. [provided by RefSeq]
Sequence: MPGSPRPAPSWVLLLRLLALLRPPGLGEACSCAPAHPQQHICHSALVIRAKISSEKVVPASADPADTEKMLRYEIKQIKMFKGFEKVKDVQYIYTPFDSSLCGVKLEANSQKQYLLTGQVLSDGKVFIHLCNYIEPWEGLSLVQRESLNHHYHLNCGCQITTCYTVPCTISAPNECLWTDWLLGRKLYGYQAQHYVCMKHVDGTCSWYRGHLPLRKEFVDIVQPSpecifications
AAH10553.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.38kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TIMP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7079 | |
TIMP4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPGSPRPAPSWVLLLRLLALLRPPGLGEACSCAPAHPQQHICHSALVIRAKISSEKVVPASADPADTEKMLRYEIKQIKMFKGFEKVKDVQYIYTPFDSSLCGVKLEANSQKQYLLTGQVLSDGKVFIHLCNYIEPWEGLSLVQRESLNHHYHLNCGCQITTCYTVPCTISAPNECLWTDWLLGRKLYGYQAQHYVCMKHVDGTCSWYRGHLPLRKEFVDIVQP | |
RUO | |
TIMP4 | |
Recombinant | |
wheat germ expression system |