missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TIMM17A Full-length ORF (NP_006326.1, 1 a.a. - 171 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010440-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195092

  • 528.00€ / 25µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

Sequence: MEEYAREPCPWRIVDDCGGAFTMGTIGGGIFQAIKGFRNSPVGVNHRLRGSLTAIKTRAPQLGGSFAVWGGLFSMIDCSMVQVRGKEDPWNSITSGALTGAILAARNGPVAMVGSAAMGGILLALIEGAGILLTRFASAQFPNGPQFAEDPSQLPSTQLPSSPFGDYRQYQ
Specifications

Specifications

NP_006326.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TIM17/TIM17A
TIMM17A
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10440
TIMM17A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEEYAREPCPWRIVDDCGGAFTMGTIGGGIFQAIKGFRNSPVGVNHRLRGSLTAIKTRAPQLGGSFAVWGGLFSMIDCSMVQVRGKEDPWNSITSGALTGAILAARNGPVAMVGSAAMGGILLALIEGAGILLTRFASAQFPNGPQFAEDPSQLPSTQLPSSPFGDYRQYQ
RUO
TIMM17A
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers