missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human THNSL1 Partial ORF (NP_079114, 151 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079896-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16160117

  • 545.00€ / 25µg

Estimated Shipment: 19-08-2022
to see stock.
Add to basket

Description

Description

Sequence: IVYLDVPLLDLICRLKLMKTDRIVGQNSGTSMKDLLKFRRQYYKKWYDARVFCESGASPEEVADKVLNAIKRYQDVDSETFISTRHVWPEDCEQKVSAKF
Specifications

Specifications

NP_079114
Liquid
79896
THNSL1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ22002/TSH1
THNSL1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IVYLDVPLLDLICRLKLMKTDRIVGQNSGTSMKDLLKFRRQYYKKWYDARVFCESGASPEEVADKVLNAIKRYQDVDSETFISTRHVWPEDCEQKVSAKF
RUO
THNSL1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers