Learn More
Abnova™ Human TH1L Full-length ORF (NP_945327.1, 1 a.a. - 590 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051497-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The NELF complex of proteins interacts with the DSIF protein complex to repress transcriptional elongation by RNA polymerase II. The protein encoded by this gene is an essential part of the NELF complex. Alternative translation initiation site usage results in the formation of two isoforms with different N-termini. [provided by RefSeq]
Sequence: MAGAVPGAIMDEDYYGSAAEWGDEADGGQQEDDSGEGEDDAEVQQECLHKFSTRDYIMEPSIFNTLKRYFQAGGSPENVIQLLSENYTAVAQTVNLLAEWLIQTGVEPVQVQETVENHLKSLLIKHFDPRKADSIFTEEGETPAWLEQMIAHTTWRDLFYKLAEAHPDCLMLNFTVKLISDAGYQGEITSVSTACQQLEVFSRVLRTSLATILDGGEENLEKNLPEFAKMVCHGEHTYLFAQAMMSVLAQEEQGGSAVRRIAQEVQRFAQEKGHDASQITLALGTAASYPRACQALGAMLSKGALNPADITVLFKMFTSMDPPPVELIRVPAFLDLFMQSLFKPGARINQDHKHKYIHILAYAASVVETWKKNKRVSINKDELKSTSKAVETVHNLCCNENKGASELVAELSTLYQCIRFPVVAMGVLKWVDWTVSEPRYFQLQTDHTPVHLALLDEISTCHQLLHPQVLQLLVKLFETEHSQLDVMEQLELKKTLLDRMVHLLSRGYVLPVVSYIRKCLEKLDTDISLIRYFVTEVLDVIAPPYTSDFVQLFLPILENDSIAGTIKTEGEHDPVTEFIAHCKSNFIMVNSpecifications
NP_945327.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
92.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSPC130/NELF-C/NELF-D/TH1 | |
TH1L | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51497 | |
TH1L (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAGAVPGAIMDEDYYGSAAEWGDEADGGQQEDDSGEGEDDAEVQQECLHKFSTRDYIMEPSIFNTLKRYFQAGGSPENVIQLLSENYTAVAQTVNLLAEWLIQTGVEPVQVQETVENHLKSLLIKHFDPRKADSIFTEEGETPAWLEQMIAHTTWRDLFYKLAEAHPDCLMLNFTVKLISDAGYQGEITSVSTACQQLEVFSRVLRTSLATILDGGEENLEKNLPEFAKMVCHGEHTYLFAQAMMSVLAQEEQGGSAVRRIAQEVQRFAQEKGHDASQITLALGTAASYPRACQALGAMLSKGALNPADITVLFKMFTSMDPPPVELIRVPAFLDLFMQSLFKPGARINQDHKHKYIHILAYAASVVETWKKNKRVSINKDELKSTSKAVETVHNLCCNENKGASELVAELSTLYQCIRFPVVAMGVLKWVDWTVSEPRYFQLQTDHTPVHLALLDEISTCHQLLHPQVLQLLVKLFETEHSQLDVMEQLELKKTLLDRMVHLLSRGYVLPVVSYIRKCLEKLDTDISLIRYFVTEVLDVIAPPYTSDFVQLFLPILENDSIAGTIKTEGEHDPVTEFIAHCKSNFIMVN | |
RUO | |
TH1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |