Learn More
R&D Systems™ Human Th1/Th2 Luminex™ Performance Assay 13-plex Fixed Panel
Product Code. 18783927
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. 18783927
Brand: R&D Systems™ LKTM008B
Specifications
Quantity 1 Kit
Species Human
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.