missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TGOLN2 Partial ORF (NP_006455, 229 a.a. - 327 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010618-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192906

  • 349.00€ / 10µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

Sequence: QGPIDGPSKSGAEEQTSKDSPNKVVPEQPSRKDHSKPISNPSDNKELPKADTNQLADKGKLSPHAFKTESGEETDLISPPQEEVKSSEPTEDVEPKEAE
Specifications

Specifications

NP_006455
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QGPIDGPSKSGAEEQTSKDSPNKVVPEQPSRKDHSKPISNPSDNKELPKADTNQLADKGKLSPHAFKTESGEETDLISPPQEEVKSSEPTEDVEPKEAE
RUO
TGOLN2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10618
TGOLN2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC14722/TGN38/TGN46/TGN48/TGN51/TTGN2
TGOLN2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers