Learn More
Abnova™ Human TFAP4 Full-length ORF (NP_003214.1, 1 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007023-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Transcription factors of the basic helix-loop-helix-zipper (bHLH-ZIP) family contain a basic domain, which is used for DNA binding, and HLH and ZIP domains, which are used for oligomerization. Transcription factor AP4 activates both viral and cellular genes by binding to the symmetrical DNA sequence CAGCTG (Mermod et al., 1988 [PubMed 2833704]; Hu et al., 1990 [PubMed 2123466]).[supplied by OMIM]
Sequence: MEYFMVPTQKVPSLQHFRKTEKEVIGGLCSLANIPLTPETQRDQERRIRREIANSNERRRMQSINAGFQSLKTLIPHTDGEKLSKAAILQQTAEYIFSLEQEKTRLLQQNTQLKRFIQELSGSSPKRRRAEDKDEGIGSPDIWEDEKAEDLRREMIELRQQLDKERSVRMMLEEQVRSLEAHMYPEKLKVIAQQVQLQQQQEQVRLLHQEKLEREQQQLRTQLLPPPAPTHHPTVIVPAPPPPPSHHINVVTMGPSSVINSVSTSRQNLDTIVQAIQHIEGTQEKQELEEEQRRAVIVKPVRSCPEAPTSDTASDSEASDSDAMDQSREEPSGDGELPSpecifications
NP_003214.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AP-4/bHLHc41 | |
TFAP4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7023 | |
TFAP4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEYFMVPTQKVPSLQHFRKTEKEVIGGLCSLANIPLTPETQRDQERRIRREIANSNERRRMQSINAGFQSLKTLIPHTDGEKLSKAAILQQTAEYIFSLEQEKTRLLQQNTQLKRFIQELSGSSPKRRRAEDKDEGIGSPDIWEDEKAEDLRREMIELRQQLDKERSVRMMLEEQVRSLEAHMYPEKLKVIAQQVQLQQQQEQVRLLHQEKLEREQQQLRTQLLPPPAPTHHPTVIVPAPPPPPSHHINVVTMGPSSVINSVSTSRQNLDTIVQAIQHIEGTQEKQELEEEQRRAVIVKPVRSCPEAPTSDTASDSEASDSDAMDQSREEPSGDGELP | |
RUO | |
TFAP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |