Learn More
R&D Systems™ Human TERT (NP_937983.2) VersaClone cDNA
Specifications
Specifications
| Gene | TERT |
| Accession Number | NP_937983.2 |
| Gene ID (Entrez) | 7015 |
| Gene Alias | EST2Telomerase-associated protein 2, hEST2, hTERT, TCS1EC 2.7.7.49, Telomerase catalytic subunit, telomerase reverse transcriptase, TP2HEST2, TRTEC 2.7.7 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |