Learn More
Abnova™ Human TEKT1 Partial ORF (NP_444515.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00083659-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. This gene is predominantly expressed in the testis and in mouse, tektin 1 mRNA was localized to the spermatocytes and round spermatids in the seminiferous tubules, indicating that it may play a role in spermatogenesis. [provided by RefSeq]
Sequence: MAKLLQPPPKFLPSEWHIANKNQYHRADAQRSRSERLVAESQRLVDEIEKTTRKSQSDVNKKLEQRLEEVQFWKKELDDKLEQLVNVTDDLLIYKIRLEKSpecifications
NP_444515.1 | |
Liquid | |
83659 | |
TEKT1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TEKT1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAKLLQPPPKFLPSEWHIANKNQYHRADAQRSRSERLVAESQRLVDEIEKTTRKSQSDVNKKLEQRLEEVQFWKKELDDKLEQLVNVTDDLLIYKIRLEK | |
RUO | |
TEKT1 | |
Yes | |
wheat germ expression system |