missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TCF7L2 Partial ORF (NP_110383, 490 a.a. - 596 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006934-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16126365

  • 335.00€ / 10µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

This gene encodes a high mobility group (HMG) box-containing transcription factor that plays a key role in the Wnt signaling pathway. The protein has been implicated in blood glucose homeostasis. Genetic variants of this gene are associated with increased risk of type 2 diabetes

Sequence: SPNLLGSPPRDAKSQTEQTQPLSLSLKPDPLAHLSMMPPPPALLLAEATHKASALCPNGALDLPPAALQPAAPSSSIAQPSTSSLHSHSSLAGTQPQPLSLVTKSLE
Specifications

Specifications

NP_110383
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.51kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SPNLLGSPPRDAKSQTEQTQPLSLSLKPDPLAHLSMMPPPPALLLAEATHKASALCPNGALDLPPAALQPAAPSSSIAQPSTSSLHSHSSLAGTQPQPLSLVTKSLE
RUO
TCF7L2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6934
TCF7L2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TCF-4/TCF4
TCF7L2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers