missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TCF12 Partial ORF (NP_996919, 364 a.a. - 453 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006938-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16106375

  • 528.00€ / 25µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of the basic helix-loop-helix (bHLH) E-protein family that recognizes the consensus binding site (E-box) CANNTG. This encoded protein is expressed in many tissues, among them skeletal muscle, thymus, B- and T-cells, and may participate in regulating lineage-specific gene expression through the formation of heterodimers with other bHLH E-proteins. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]

Sequence: VGSPSPLTGTSQWPRPGGQAPSSPSYENSLHSLKNRVEQQLHEHLQDAMSFLKDVCEQSRMEDRLDRLDDAIHVLRNHAVGPSTSLPAGH
Specifications

Specifications

NP_996919
Liquid
6938
TCF12 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HEB/HTF4/HsT17266/bHLHb20
TCF12
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VGSPSPLTGTSQWPRPGGQAPSSPSYENSLHSLKNRVEQQLHEHLQDAMSFLKDVCEQSRMEDRLDRLDDAIHVLRNHAVGPSTSLPAGH
RUO
TCF12
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers