missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TCAP Full-length ORF (AAH12628, 1 a.a. - 167 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008557-P02.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192572

  • 528.00€ / 25µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. This gene encodes a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in this gene are associated with limb-girdle muscular dystrophy type 2G. [provided by RefSeq]

Sequence: MATSELSCEVSEENCERREAFWAEWKDLTLSTRPEEGCSLHEEDTQRHETYHQQGQCQVLVQRSPWLMMRMGILGRGLQEYQLPYQRVLPLPIFTPAKMGATKEEREDTPIQLQELLALETALGGQCVDRQEVAEITKQLPPVVPVSKPGALRRSLSRSMSQEAQRG
Specifications

Specifications

AAH12628
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.11kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CMD1N/LGMD2G/T-cap/TELE/telethonin
TCAP
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8557
TCAP (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATSELSCEVSEENCERREAFWAEWKDLTLSTRPEEGCSLHEEDTQRHETYHQQGQCQVLVQRSPWLMMRMGILGRGLQEYQLPYQRVLPLPIFTPAKMGATKEEREDTPIQLQELLALETALGGQCVDRQEVAEITKQLPPVVPVSKPGALRRSLSRSMSQEAQRG
RUO
TCAP
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers