missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TBCE Full-length ORF (NP_003184.1, 1 a.a. - 527 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006905-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123428

  • 335.00€ / 10µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

Cofactor E is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: MSDTLTADVIGRRVEVNGEHATVRFAGVVPPVAGPWLGVEWDNPERGKHDGSHEGTVYFKCRHPTGGSFIRPNKVNFGTDFLTAIKNRYVLEDGPEEDRKEQIVTIGNKPVETIGFDSIMKQQSQLSKLQEVSLRNCAVSCAGEKGGVAEACPNIRKVDLSKNLLSSWDEVIHIADQLRHLEVLNVSENKLKFPSGSVLTGTLSVLKVLVLNQTGITWAEVLRCVAGCPGLEELYLESNNIFISERPTDVLQTVKLLDLSSNQLIDENQLYLIAHLPRLEQLILSDTGISSLHFPDAGIGCKTSMFPSLKYLVVNDNQISQWSFFNELEKLPSLRALSCLRNPLTKEDKEAETARLLIIASIGQLKTLNKCEILPEERRRAELDYRKAFGNEWKQAGGHKDPEKNRLSEEFLTAHPRYQFLCLKYGAPEDWELKTQQPLMLKNQLLTLKIKYPHQLDQKVLEKQLPGSMTIQKVKGLLSRLLKVPVSDLLLSYESPKKPGREIELENDLKSLQFYSVENGDCLLVRW
Specifications

Specifications

NP_003184.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
85.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HRD/KCS/KCS1/pac2
TBCE
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
6905
TBCE (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSDTLTADVIGRRVEVNGEHATVRFAGVVPPVAGPWLGVEWDNPERGKHDGSHEGTVYFKCRHPTGGSFIRPNKVNFGTDFLTAIKNRYVLEDGPEEDRKEQIVTIGNKPVETIGFDSIMKQQSQLSKLQEVSLRNCAVSCAGEKGGVAEACPNIRKVDLSKNLLSSWDEVIHIADQLRHLEVLNVSENKLKFPSGSVLTGTLSVLKVLVLNQTGITWAEVLRCVAGCPGLEELYLESNNIFISERPTDVLQTVKLLDLSSNQLIDENQLYLIAHLPRLEQLILSDTGISSLHFPDAGIGCKTSMFPSLKYLVVNDNQISQWSFFNELEKLPSLRALSCLRNPLTKEDKEAETARLLIIASIGQLKTLNKCEILPEERRRAELDYRKAFGNEWKQAGGHKDPEKNRLSEEFLTAHPRYQFLCLKYGAPEDWELKTQQPLMLKNQLLTLKIKYPHQLDQKVLEKQLPGSMTIQKVKGLLSRLLKVPVSDLLLSYESPKKPGREIELENDLKSLQFYSVENGDCLLVRW
RUO
TBCE
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers