missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TBCD Full-length ORF (AAH39654.1, 1 a.a. - 623 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH39654.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6904 |
Molecular Weight (g/mol) | 95.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16113428
|
Abnova™
H00006904-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16103428
|
Abnova™
H00006904-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
Cofactor D is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state. [provided by RefSeq]
Sequence: MKISHWDGVIRELAARALHNLAQQAPEFSATQVFPRLLSMTLSPDLHTRHGSILACAEVAYALYKLAAQENRPVTDHLDEQAVQGLKQIHQQLYDRQLYRGLGGQLMRQAVCVLIEKLSLSKMPFRGDTVIDGWQWLINDTLRHLHLISSHSRQQMKDAAVSALAALCSEYYMKEPGEADPAIQEELITQYLAELRNPEEMTRCGFSLALGALPGFLLKGRLQQVLTGLRAVTHTSPEDVSFAESRRDGLKAIARICQTVGVKAGAPDEAVCGENVSQIYCALLGCMDDYTTDSRGDVGTWVRKAAMTSLMDLTLLLARSQPELIEAHTCERIMCCVAQQASEKIDRFRAHAASVFLTLLHFDSPPIPHVPHRGELEKLFPRSDVASVNWSAPSQAFPRITQLLGLPTYRYHVLLGLVVSLGGLTESTIRHSTQSLFEYMKGIQSDPQALGSFSGTLLQIFEDNLLNERVSVPLLKTLDHVLTHGCFDIFTTEEDHPFAVKLLALCKKEIKNSKDIQKLLSGIAVFCGMVQFPGDVRRQALLQLCLLLCHRFPLIRKTTASQVYETLLTYSDVVGADVLDEVVTVLSDTAWDAELAVVREQRNRLCDLLGVPRPQLVPQPGACSpecifications
AAH39654.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
95.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKISHWDGVIRELAARALHNLAQQAPEFSATQVFPRLLSMTLSPDLHTRHGSILACAEVAYALYKLAAQENRPVTDHLDEQAVQGLKQIHQQLYDRQLYRGLGGQLMRQAVCVLIEKLSLSKMPFRGDTVIDGWQWLINDTLRHLHLISSHSRQQMKDAAVSALAALCSEYYMKEPGEADPAIQEELITQYLAELRNPEEMTRCGFSLALGALPGFLLKGRLQQVLTGLRAVTHTSPEDVSFAESRRDGLKAIARICQTVGVKAGAPDEAVCGENVSQIYCALLGCMDDYTTDSRGDVGTWVRKAAMTSLMDLTLLLARSQPELIEAHTCERIMCCVAQQASEKIDRFRAHAASVFLTLLHFDSPPIPHVPHRGELEKLFPRSDVASVNWSAPSQAFPRITQLLGLPTYRYHVLLGLVVSLGGLTESTIRHSTQSLFEYMKGIQSDPQALGSFSGTLLQIFEDNLLNERVSVPLLKTLDHVLTHGCFDIFTTEEDHPFAVKLLALCKKEIKNSKDIQKLLSGIAVFCGMVQFPGDVRRQALLQLCLLLCHRFPLIRKTTASQVYETLLTYSDVVGADVLDEVVTVLSDTAWDAELAVVREQRNRLCDLLGVPRPQLVPQPGAC | |
RUO | |
TBCD | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
6904 | |
TBCD (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0988 | |
TBCD | |
Recombinant | |
wheat germ expression system |