missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TAS2R60 Full-length ORF (NP_803186.1, 1 a.a. - 318 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00338398-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16110124

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MNGDHMVLGSSVTDKKAIILVTILLLLRLVAIAGNGFITAALGVEWVLRRMLLPCDKLLVSLGASRFCLQSVVMGKTIYVFLHPMAFPYNPVLQFLAFQWDFLNAATLWSSTWLSVFYCVKIATFTHPVFFWLKHKLSGWLPWMLFSSVGLSSFTTILFFIGNHRMYQNYLRNHLQPWNVTGDSIRSYCEKFYLFPLKMITWTMPTAVFFICMILLITSLGRHRKKALLTTSGFREPSVQAHIKALLALLSFAMLFISYFLSLVFSAAGIFPPLDFKFWVWESVIYLCAAVHPIILLFSNCRLRAVLKSRRSSRCGTP
Specifications

Specifications

NP_803186.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC119154/MGC119155/T2R56/T2R60
TAS2R60
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
338398
TAS2R60 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNGDHMVLGSSVTDKKAIILVTILLLLRLVAIAGNGFITAALGVEWVLRRMLLPCDKLLVSLGASRFCLQSVVMGKTIYVFLHPMAFPYNPVLQFLAFQWDFLNAATLWSSTWLSVFYCVKIATFTHPVFFWLKHKLSGWLPWMLFSSVGLSSFTTILFFIGNHRMYQNYLRNHLQPWNVTGDSIRSYCEKFYLFPLKMITWTMPTAVFFICMILLITSLGRHRKKALLTTSGFREPSVQAHIKALLALLSFAMLFISYFLSLVFSAAGIFPPLDFKFWVWESVIYLCAAVHPIILLFSNCRLRAVLKSRRSSRCGTP
RUO
TAS2R60
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers