Learn More
Abnova™ Human TAS2R43 Full-length ORF (AAI17424.1, 1 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00259289-P01.25ug
Additional Details : Weight : 0.00010kg
Description
TAS2R43 belongs to the large TAS2R receptor family. TAS2Rs are expressed on the surface of taste receptor cells and mediate the perception of bitterness through a G protein-coupled second messenger pathway (Conte et al., 2002 [PubMed 12584440]). For further information on TAS2Rs, see MIM 604791.[supplied by OMIM]
Sequence: MITFLPIIFSSLVVVTFVIGNFANGFIALVNSIESFKRQKISFADQILTALAVSRVGLLWVLLLNWYSTVLNPAFNSVEVRTTAYNIWAVINHFSNWLATTLSIFYLLKIANFSNFIFLHLKRRVKSVILVMLLGPLLFLACHLFVINMNEIVRTKEFEGNMTWKIKLKSAMYFSNMTVTMVANLVPFTLTLLSFMLLICSLCKHLKKMQLRGKGSQDPSTKVHIKALQTVISFLLLCAIYFLSIMISVWSFGSLENKPVFMFCKAIRFSYPSIHPFILIWGNKKLKQTFLSVFWQMRYWVKGEKTSSPSpecifications
AAI17424.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
T2R43/T2R52 | |
TAS2R43 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
259289 | |
TAS2R43 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MITFLPIIFSSLVVVTFVIGNFANGFIALVNSIESFKRQKISFADQILTALAVSRVGLLWVLLLNWYSTVLNPAFNSVEVRTTAYNIWAVINHFSNWLATTLSIFYLLKIANFSNFIFLHLKRRVKSVILVMLLGPLLFLACHLFVINMNEIVRTKEFEGNMTWKIKLKSAMYFSNMTVTMVANLVPFTLTLLSFMLLICSLCKHLKKMQLRGKGSQDPSTKVHIKALQTVISFLLLCAIYFLSIMISVWSFGSLENKPVFMFCKAIRFSYPSIHPFILIWGNKKLKQTFLSVFWQMRYWVKGEKTSSP | |
RUO | |
TAS2R43 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |