missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TAIP-2 Partial ORF (NP_079245, 476 a.a. - 585 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00080034-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194743

  • 528.00€ / 25µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

Sequence: QFVDYARQAEEAYGASHYPAANPSVIVCCSSSENDSGVPCNSLYPEHRSNHPQVEFHSYLKGPSQEGFVSALNGDSHISEHPAENSLSLAEKSILHEECIKSPVVETVPV
Specifications

Specifications

NP_079245
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QFVDYARQAEEAYGASHYPAANPSVIVCCSSSENDSGVPCNSLYPEHRSNHPQVEFHSYLKGPSQEGFVSALNGDSHISEHPAENSLSLAEKSILHEECIKSPVVETVPV
RUO
CSRNP3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
80034
TAIP-2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FAM130A2/FLJ11703/FLJ32093/FLJ44141/TAIP-2/TAIP2
CSRNP3
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers