Learn More
Abnova™ Human TAF5 Partial ORF (NP_008882, 428 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006877-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes an integral subunit of TFIID associated with all transcriptionally competent forms of that complex. This subunit interacts strongly with two TFIID subunits that show similarity to histones H3 and H4, and it may participate in forming a nucleosome-like core in the TFIID complex. [provided by RefSeq]
Sequence: PLPELKDSDKLDKIMNMKETTKRVRLGPDCLPSICFYTFLNAYQGLTAVDVTDDSSLIAGGFADSTVRVWSVTPKKLRSVKQASDLSLIDKESDDVLERIMDEKTASESpecifications
NP_008882 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PLPELKDSDKLDKIMNMKETTKRVRLGPDCLPSICFYTFLNAYQGLTAVDVTDDSSLIAGGFADSTVRVWSVTPKKLRSVKQASDLSLIDKESDDVLERIMDEKTASE | |
RUO | |
TAF5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6877 | |
TAF5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TAF2D/TAFII100 | |
TAF5 | |
Recombinant | |
wheat germ expression system |