Learn More
Abnova™ Human T Full-length ORF (AAH98425.1, 1 a.a. - 377 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006862-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells. [provided by RefSeq]
Sequence: MSSPGTESAGKSLQYRVDHLLSAVENELQAGSEKGDPTERELRVGLEESELWLRFKELTNEMIVTKNGRRMFPVLKVNVSGLDPNAMYSFLLDFVAADNHRWKYVNGEWVPGGKPEPQAPSCVYIHPDSPNFGAHWMKAPVSFSKVKLTNKLNGGGQIMLNSLHKYEPRIHIVRVGDPQRMITSHCFPETQFIAVTAYQNEEITALKIKYNPFAKAFLDAKERSDHKEMMEEPGDSQQPGYSQSYSDNSPACLSMLQSHDNWSSLGMPAHPSMLPVSHNASPPTSSSQYPSLWSVSNGAVTPGSQAAAVSNGLGAQFFRGSPAHYTPLTHPVSAPSSSGSPLYEGAAAATDIVDSQYDAAAQGRLIASWTPVSPPSMSpecifications
AAH98425.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC104817/TFT | |
T | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6862 | |
T (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSPGTESAGKSLQYRVDHLLSAVENELQAGSEKGDPTERELRVGLEESELWLRFKELTNEMIVTKNGRRMFPVLKVNVSGLDPNAMYSFLLDFVAADNHRWKYVNGEWVPGGKPEPQAPSCVYIHPDSPNFGAHWMKAPVSFSKVKLTNKLNGGGQIMLNSLHKYEPRIHIVRVGDPQRMITSHCFPETQFIAVTAYQNEEITALKIKYNPFAKAFLDAKERSDHKEMMEEPGDSQQPGYSQSYSDNSPACLSMLQSHDNWSSLGMPAHPSMLPVSHNASPPTSSSQYPSLWSVSNGAVTPGSQAAAVSNGLGAQFFRGSPAHYTPLTHPVSAPSSSGSPLYEGAAAATDIVDSQYDAAAQGRLIASWTPVSPPSM | |
RUO | |
T | |
Wheat Germ (in vitro) | |
GST | |
Liquid |