missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SYT2 Partial ORF (NP_796376, 311 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00127833-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101747

  • 349.00€ / 10µg

Estimated Shipment: 17-07-2023
to see stock.
Add to basket

Description

Description

Synaptotagmins, like SYT2, are integral membrane proteins of synaptic vesicles thought to serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis (Hilbush and Morgan, 1994 [PubMed 8058779]).[supplied by OMIM]

Sequence: KIHLMQNGKRLKKKKTTVKKKTLNPYFNESFSFEIPFEQIQKVQVVVTVLDYDKLGKNEAIGKIFVGSNATGTELRHWSDMLANPRRPIAQWHSLKPEEEVDALLGKN
Specifications

Specifications

NP_796376
Liquid
127833
SYT2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SytII
SYT2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KIHLMQNGKRLKKKKTTVKKKTLNPYFNESFSFEIPFEQIQKVQVVVTVLDYDKLGKNEAIGKIFVGSNATGTELRHWSDMLANPRRPIAQWHSLKPEEEVDALLGKN
RUO
SYT2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers