Learn More
Abnova™ Human STK31 Full-length ORF (AAH59374.1, 1 a.a. - 1019 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00056164-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is similar to a mouse gene that encodes a putative protein kinase with a tudor domain, and shows testis-specific expression. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MWVQGHSSRASATESVSFSGIVQMDEDTHYDKVEDVVGSHIEDAVTFWAQSINRNKDIMKIGCSLSEVCPHASSVLGNLDPNKIYGGLFSEDQCWYRCKVLKIISVEKCLVRYIDYGNTEILNRSDIVEIPLELQFSSVAKKYKLWGLHIPSDQEVTQFDQGTTFLGSLIFEKEIKMRIKATSEDGTVIAQAEYGSVDIGEEVLKKGFAEKCRLASRTDICEEKKLDPGQLVLRNLKSPIPLWGHRSNQSTFSRPKGHLSEKMTLDLKDENDAGNLITFPKESLAVGDFNLGSNVSLEKIKQDQKLIEENEKLKTEKDALLESYKALELKVEQIAQELQQEKAAAVDLTNHLEYTLKTYIDTRMKNLAAKMEILKEMRHVDISVRFGKDLSDAIQVLDEGCFTTPASLNGLEIIWAEYSLAQENIKTCEYVSEGNILIAQRNEMQQKLYMSVEDFILEVDESSLNKRLKTLQDLSVSLEAVYGQAKEGANSDEILKKFYDWKCDKREEFTSVRSETDASLHRLVAWFQRTLKVFDLSVEGSLISEDAMDNIDEILEKTESSVCKELEIALVDQGDADKEIISNTYSQVLQKIHSEERLIATVQAKYKDSIEFKKQLIEYLNKSPSVDHLLSIKKTLKSLKALLRWKLVEKSNLEESDDPDGSQIEKIKEEITQLRNNVFQEIYHEREEYEMLTSLAQKWFPELPLLHPEIGLLKYMNSGGLLTMSLERDLLDAEPMKELSSKRPLVRSEVNGQIILLKGYSVDVDTEAKVIERAATYHRAWREAEGDSGLLPLIFLFLCKSDPMAYLMVPYYPRANLNAVQANMPLNSEETLKVMKGVAQGLHTLHKADIIHGSLHQNNVFALNREQGIVGDFDFTKSVSQRASVNMMVGDLSLMSPELKMGKPASPGSDLYAYGCLLLWLSVQNQEFEINKDGIPKVDQFHLDDKVKSLLCSLICYRSSMTAEQVLNAECFLMPKEQSVPNPEKDTEYTLYKKEEEIKTENLDKCMEKTRNGEANFDCSpecifications
AAH59374.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
142.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ16102/SGK396/TDRD8 | |
STK31 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
56164 | |
STK31 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MWVQGHSSRASATESVSFSGIVQMDEDTHYDKVEDVVGSHIEDAVTFWAQSINRNKDIMKIGCSLSEVCPHASSVLGNLDPNKIYGGLFSEDQCWYRCKVLKIISVEKCLVRYIDYGNTEILNRSDIVEIPLELQFSSVAKKYKLWGLHIPSDQEVTQFDQGTTFLGSLIFEKEIKMRIKATSEDGTVIAQAEYGSVDIGEEVLKKGFAEKCRLASRTDICEEKKLDPGQLVLRNLKSPIPLWGHRSNQSTFSRPKGHLSEKMTLDLKDENDAGNLITFPKESLAVGDFNLGSNVSLEKIKQDQKLIEENEKLKTEKDALLESYKALELKVEQIAQELQQEKAAAVDLTNHLEYTLKTYIDTRMKNLAAKMEILKEMRHVDISVRFGKDLSDAIQVLDEGCFTTPASLNGLEIIWAEYSLAQENIKTCEYVSEGNILIAQRNEMQQKLYMSVEDFILEVDESSLNKRLKTLQDLSVSLEAVYGQAKEGANSDEILKKFYDWKCDKREEFTSVRSETDASLHRLVAWFQRTLKVFDLSVEGSLISEDAMDNIDEILEKTESSVCKELEIALVDQGDADKEIISNTYSQVLQKIHSEERLIATVQAKYKDSIEFKKQLIEYLNKSPSVDHLLSIKKTLKSLKALLRWKLVEKSNLEESDDPDGSQIEKIKEEITQLRNNVFQEIYHEREEYEMLTSLAQKWFPELPLLHPEIGLLKYMNSGGLLTMSLERDLLDAEPMKELSSKRPLVRSEVNGQIILLKGYSVDVDTEAKVIERAATYHRAWREAEGDSGLLPLIFLFLCKSDPMAYLMVPYYPRANLNAVQANMPLNSEETLKVMKGVAQGLHTLHKADIIHGSLHQNNVFALNREQGIVGDFDFTKSVSQRASVNMMVGDLSLMSPELKMGKPASPGSDLYAYGCLLLWLSVQNQEFEINKDGIPKVDQFHLDDKVKSLLCSLICYRSSMTAEQVLNAECFLMPKEQSVPNPEKDTEYTLYKKEEEIKT | |
RUO | |
STK31 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |