missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human STIP1 Partial ORF (AAH02987.1, 320 a.a. - 464 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010963-Q06.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16015295

  • 508.00€ / 25µg
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

STIP1 is an adaptor protein that coordinates the functions of HSP70 (see HSPA1A; MIM 140550) and HSP90 (see HSP90AA1; MIM 140571) in protein folding. It is thought to assist in the transfer of proteins from HSP70 to HSP90 by binding both HSP90 and substrate-bound HSP70. STIP1 also stimulates the ATPase activity of HSP70 and inhibits the ATPase activity of HSP90, suggesting that it regulates both the conformations and ATPase cycles of these chaperones (Song and Masison, 2005 [PubMed 16100115]).[supplied by OMIM]

Sequence: IHFYNKSLAEHRTPDVLKKCQQAEKILKEQERLAYINPDLALEEKNKGNECFQKGDYPQAMKHYTEAIKRNPKDAKLYSNRAACYTKLLEFQLALKDCEECIQLEPTFIKGYTRKAAALEAMKDYTKAMDVYQKALDLDSSCKEA
Specifications

Specifications

AAH02987.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
41.58kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IHFYNKSLAEHRTPDVLKKCQQAEKILKEQERLAYINPDLALEEKNKGNECFQKGDYPQAMKHYTEAIKRNPKDAKLYSNRAACYTKLLEFQLALKDCEECIQLEPTFIKGYTRKAAALEAMKDYTKAMDVYQKALDLDSSCKEA
RUO
STIP1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10963
STIP1 (Human) Recombinant Protein (Q06)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HOP/IEF-SSP-3521/P60/STI1/STI1L
STIP1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers