missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human STARD8 Full-length ORF (NP_055540.2, 1 a.a. - 1023 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009754-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MTLNNCASMKLEVHFQSKQNEDSEEEEQCTISSHWAFQQESKCWSPMGSSDLLAPPSPGLPATSSCESVLTELSATSLPVITVSLPPEPADLPLPGRAPSSSDRPLLSPTQGQEGPQDKAKKRHRNRSFLKHLESLRRKEKSGSQQAEPKHSPATSEKVSKASSFRSCRGFLSAGFYRAKNWAATSAGGSGANTRKAWEAWPVASFRHPQWTHRGDCLVHVPGDHKPGTFPRSLSIESLCPEDGHRLADWQPGRRWGCEGRRGSCGSTGSHASTYDNLPELYPAEPVMVGAEAEDEDDEESGGSYAHLDDILQHVWGLQQRVELWSRAMYPDLGPGDEEEEEATSSVEIATVEVKCQAEALSQMEVPAHGESPAWAQAEVQPAVLAPAQAPAEAEPVAQEEAEAPAPAPAPAPAQDSEQEAHSGGEPTFASSLSVEEGHSISDTVASSSELDSSGNSMNEAEAAGPLAGLQASMPRERRDSGVGASLTRPCRKLRWHSFQNSHRPSLNSESLEINRQFAGQINLLHKGSLLRLTAFMEKYTVPHKQGWVWSMPKFMRRNKTPDYRGQHVFGVPPLIHVQRTGQPLPQSIQQAMRYLRSQCLDQVGIFRKSGVKSRIQNLRQMNETSPDNVCYEGQSAYDVADLLKQYFRDLPEPIFTSKLTTTFLQIYQLLPKDQWLAAAQAATLLLPDENREVLQTLLYFLSDIASAEENQMTAGNLAVCLAPSIFHLNVSKKDSPSPRIKSKRSLIGRPGPRDLSDNMAATQGLSHMISDCKKLFQVPQDMVLQLCSSYSAAELSPPGPALAELRQAQAAGVSLSLYMEENIQDLLRDAAERFKGWMSVPGPQHTELACRKAPDGHPLRLWKASTEVAAPPAVVLHRVLRERALWDEDLLRAQVLEALMPGVELYHYVTDSMAPHPCRDFVVLRMWRSDLPRGGCLLVSQSLDPEQPVPESGVRALMLTSQYLMEPCGLGRSRLTHICRADLRGRSPDWYNKVFGHLCAMEVAKIRDSFPTLQAAGPETKLSpecifications
NP_055540.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
139kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686H1668/DLC3/KIAA0189/STARTGAP3 | |
STARD8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9754 | |
STARD8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTLNNCASMKLEVHFQSKQNEDSEEEEQCTISSHWAFQQESKCWSPMGSSDLLAPPSPGLPATSSCESVLTELSATSLPVITVSLPPEPADLPLPGRAPSSSDRPLLSPTQGQEGPQDKAKKRHRNRSFLKHLESLRRKEKSGSQQAEPKHSPATSEKVSKASSFRSCRGFLSAGFYRAKNWAATSAGGSGANTRKAWEAWPVASFRHPQWTHRGDCLVHVPGDHKPGTFPRSLSIESLCPEDGHRLADWQPGRRWGCEGRRGSCGSTGSHASTYDNLPELYPAEPVMVGAEAEDEDDEESGGSYAHLDDILQHVWGLQQRVELWSRAMYPDLGPGDEEEEEATSSVEIATVEVKCQAEALSQMEVPAHGESPAWAQAEVQPAVLAPAQAPAEAEPVAQEEAEAPAPAPAPAPAQDSEQEAHSGGEPTFASSLSVEEGHSISDTVASSSELDSSGNSMNEAEAAGPLAGLQASMPRERRDSGVGASLTRPCRKLRWHSFQNSHRPSLNSESLEINRQFAGQINLLHKGSLLRLTAFMEKYTVPHKQGWVWSMPKFMRRNKTPDYRGQHVFGVPPLIHVQRTGQPLPQSIQQAMRYLRSQCLDQVGIFRKSGVKSRIQNLRQMNETSPDNVCYEGQSAYDVADLLKQYFRDLPEPIFTSKLTTTFLQIYQLLPKDQWLAAAQAATLLLPDENREVLQTLLYFLSDIASAEENQMTAGNLAVCLAPSIFHLNVSKKDSPSPRIKSKRSLIGRPGPRDLSDNMAATQGLSHMISDCKKLFQVPQDMVLQLCSSYSAAELSPPGPALAELRQAQAAGVSLSLYMEENIQDLLRDAAERFKGWMSVPGPQHTELACRKAPDGHPLRLWKASTEVAAPPAVVLHRVLRERALWDEDLLRAQVLEALMPGVELYHYVTDSMAPHPCRDFVVLRMWRSDLPRGGCLLVSQSLDPEQPVPESGVRALMLTSQYLMEPCGLGRSRLTHICRADLRGRSPDWYNKVFGHLC | |
RUO | |
STARD8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |