missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human STARD10 (NP_006636.2) VersaClone cDNA
Specifications
Specifications
| Gene | STARD10 |
| Accession Number | NP_006636.2 |
| Gene ID (Entrez) | 10809 |
| Gene Alias | Antigen NY-CO-28, CGI-52, MGC14401, NY-CO-28, PCTP2, PCTP-L, PCTP-like protein, SDCCAG28, Serologically defined colon cancer antigen 28 SDCCAG28, StARD10, StAR-related lipid transfer (START) domain containing 10, StAR-related lipid transfer protein 10, START domain containing 10, START domain-containing protein 10 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?