missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ST7L Full-length ORF (NP_060214.2, 1 a.a. - 575 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054879-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16160853

  • 359.00€ / 10µg

Estimated Shipment: 22-08-2022
to see stock.
Add to basket

Description

Description

This gene was identified by its similarity to the ST7 tumor suppressor gene found in the chromosome 7q31 region. This gene is clustered in a tail-to-tail manner with the WNT2B gene in a chromosomal region known to be deleted and rearranged in a variety of cancers. The function of this gene has not yet been determined. Several transcript variants encoding many different isoforms have been described, but some have not been fully characterized. [provided by RefSeq]

Sequence: MADRGGVGEAAAVGASPASVPGLNPTLGWRERLRAGLAGTGASLWFVAGLGLLYALRIPLRLCENLAAVTVFLNSLTPKFYVALTGTSSLISGLIFIFEWWYFHKHGTSFIEQVSVSHLQPLMGGTESSISEPGSPSRNRENETSRQNLSECKVWRNPLNLFRGAEYRRYTWVTGKEPLTYYDMNLSAQDHQTFFTCDTDFLRPSDTVMQKAWRERNPPARIKAAYQALELNNDCATAYVLLAEEEATTIVDAERLFKQALKAGETIYRQSQQCQHQSPQHEAQLRRDTNVLVYIKRRLAMCARKLGRIREAVKIMRDLMKEFPPLTMLNIHENLLESLLELQAYPDVQAVLAKYDDISLPKSAAICYTAALLKTRTVSEKFSPETASRRGLSTAEINAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLQHWKRIEGALNLLQCTWEGTFRMIPYPLEKGHLFYPYPSCTETADRELLPTFHHVSVYPKKELPLFIHFTAGFCSSTAMIAILTHQFPEIMGIFAKAVLGLWCPQPWASSGFEENTQDLKSEDLGLSSG
Specifications

Specifications

NP_060214.2
Liquid
54879
ST7L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MADRGGVGEAAAVGASPASVPGLNPTLGWRERLRAGLAGTGASLWFVAGLGLLYALRIPLRLCENLAAVTVFLNSLTPKFYVALTGTSSLISGLIFIFEWWYFHKHGTSFIEQVSVSHLQPLMGGTESSISEPGSPSRNRENETSRQNLSECKVWRNPLNLFRGAEYRRYTWVTGKEPLTYYDMNLSAQDHQTFFTCDTDFLRPSDTVMQKAWRERNPPARIKAAYQALELNNDCATAYVLLAEEEATTIVDAERLFKQALKAGETIYRQSQQCQHQSPQHEAQLRRDTNVLVYIKRRLAMCARKLGRIREAVKIMRDLMKEFPPLTMLNIHENLLESLLELQAYPDVQAVLAKYDDISLPKSAAICYTAALLKTRTVSEKFSPETASRRGLSTAEINAVEAIHRAVEFNPHVPKYLLEMKSLILPPEHILKRGDSEAIAYAFFHLQHWKRIEGALNLLQCTWEGTFRMIPYPLEKGHLFYPYPSCTETADRELLPTFHHVSVYPKKELPLFIHFTAGFCSSTAMIAILTHQFPEIMGIFAKAVLGLWCPQPWASSGFEENTQDLKSEDLGLSSG
RUO
ST7L
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
91.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FAM4B/FLJ11657/FLJ20284/ST7R/STLR
ST7L
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers