missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ST3GAL5 Full-length ORF (AAH65936.1, 1 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008869-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16103022

  • 528.00€ / 25µg

Estimated Shipment: 04-07-2023
to see stock.
Add to basket

Description

Description

Ganglioside GM3 is known to participate in the induction of cell differentiation, modulation of cell proliferation, maintenance of fibroblast morphology, signal transduction, and integrin-mediated cell adhesion. The protein encoded by this gene is a type II membrane protein which catalyzes the formation of GM3 using lactosylceramide as the substrate. The encoded protein is a member of glycosyltransferase family 29 and may be localized to the Golgi apparatus. Mutation in this gene has been associated with Amish infantile epilepsy syndrome. Transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MPSEYTYVKLRSDCSRPSLQWYTRAQSKMRRPSLLLKDILKCTLLVFGVWILYILKLNYTTEECDMKKMHYVDPDRVKRAQKYAQQVLQKECRPKFAKTSMALLFEHRYSVDLLPFVQKAPKDSEAESKYDPPFGFRKFSSKVQTLLELLPEHDLPEHLKAKTCRRCVVIGSGGILHGLELGHTLNQFDVVIRLNSAPVEGYSEHVGNKTTIRMTYPEGAPLSDLEYYSNDLFVAVLFKSVDFNWLQAMVKKETLPFWVRLFFWKQVAEKIPLQPKHFRILNPVIIKETAFDILQYSEPQSRFWGRDKNVPTIGVIAVVLATHLCDEVSLAGFGYDLNQPRTPLHYFDSQCMAAMNFQTMHNVTTETKFLLKLVKEGVVKDLSGGIDREF
Specifications

Specifications

AAH65936.1
Liquid
8869
ST3GAL5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPSEYTYVKLRSDCSRPSLQWYTRAQSKMRRPSLLLKDILKCTLLVFGVWILYILKLNYTTEECDMKKMHYVDPDRVKRAQKYAQQVLQKECRPKFAKTSMALLFEHRYSVDLLPFVQKAPKDSEAESKYDPPFGFRKFSSKVQTLLELLPEHDLPEHLKAKTCRRCVVIGSGGILHGLELGHTLNQFDVVIRLNSAPVEGYSEHVGNKTTIRMTYPEGAPLSDLEYYSNDLFVAVLFKSVDFNWLQAMVKKETLPFWVRLFFWKQVAEKIPLQPKHFRILNPVIIKETAFDILQYSEPQSRFWGRDKNVPTIGVIAVVLATHLCDEVSLAGFGYDLNQPRTPLHYFDSQCMAAMNFQTMHNVTTETKFLLKLVKEGVVKDLSGGIDREF
RUO
ST3GAL5
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
71.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SIAT9/SIATGM3S/ST3GalV
ST3GAL5
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers