missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SSTR1 Partial ORF (AAH35618, 1 a.a. - 60 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006751-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16115915

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The biological effects of somatostatin are probably mediated by a family of G protein-coupled receptors that are expressed in a tissue-specific manner. The encoded protein is a member of the superfamily of somatostatin receptors having seven transmembrane segments, and is expressed in highest levels in jejunum and stomach. [provided by RefSeq]

Sequence: MFPNGTASSPSSSPSPSPGSCGEGGGSRGPGAGAADGMEEPGRNASQNGTLSEGQGSAIL
Specifications

Specifications

AAH35618
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.23kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MFPNGTASSPSSSPSPSPGSCGEGGGSRGPGAGAADGMEEPGRNASQNGTLSEGQGSAIL
RUO
SSTR1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6751
SSTR1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SRIF-2
SSTR1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers