Learn More
Abnova™ Human SSH1 Partial ORF (NP_061857.2, 950 a.a. - 1049 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054434-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The ADF (actin-depolymerizing factor)/cofilin family (see MIM 601442) is composed of stimulus-responsive mediators of actin dynamics. ADF/cofilin proteins are inactivated by kinases such as LIM domain kinase-1 (LIMK1; MIM 601329). The SSH family appears to play a role in actin dynamics by reactivating ADF/cofilin proteins in vivo (Niwa et al., 2002 [PubMed 11832213]).[supplied by OMIM]
Sequence: LVKQRTQEIETRLRLAGLTVSSPLKRSHSLAKLGSLTFSTEDLSSEADPSTVADSQDTTLSESSFLHEPQGTPRDPAATSKPSGKPAPENLKSPSWMSKSSpecifications
NP_061857.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LVKQRTQEIETRLRLAGLTVSSPLKRSHSLAKLGSLTFSTEDLSSEADPSTVADSQDTTLSESSFLHEPQGTPRDPAATSKPSGKPAPENLKSPSWMSKS | |
RUO | |
SSH1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54434 | |
SSH1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ38102/KIAA1298/SSH-1 | |
SSH1 | |
Recombinant | |
wheat germ expression system |