Learn More
Abnova™ Human SRM Full-length ORF (NP_003123.2, 1 a.a. - 302 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006723-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The polyamines putrescine, spermine, and spermidine are ubiquitous polycationic mediators of cell growth and differentiation. Spermidine synthase is one of four enzymes in the polyamine-biosynthetic pathway and carries out the final step of spermidine biosynthesis. This enzyme catalyzes the conversion of putrescine to spermidine using decarboxylated S-adenosylmethionine as the cofactor. [provided by RefSeq]
Sequence: MEPGPDGPAASGPAAIREGWFRETCSLWPGQALSLQVEQLLHHRRSRYQDILVFRSKTYGNVLVLDGVIQCTERDEFSYQEMIANLPLCSHPNPRKVLIIGGGDGGVLREVVKHPSVESVVQCEIDEDVIQVSKKFLPGMAIGYSSSKLTLHVGDGFEFMKQNQDAFDVIITDSSDPMGPAESLFKESYYQLMKTALKEDGVLCCQGECQWLHLDLIKEMRQFCQSLFPVVAYAYCTIPTYPSGQIGFMLCSKNPSTNFQEPVQPLTQQQVAQMQLKYYNSDVHRAAFVLPEFARKALNDVSSpecifications
NP_003123.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PAPT/SPDSY/SPS1/SRML1 | |
SRM | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6723 | |
SRM (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEPGPDGPAASGPAAIREGWFRETCSLWPGQALSLQVEQLLHHRRSRYQDILVFRSKTYGNVLVLDGVIQCTERDEFSYQEMIANLPLCSHPNPRKVLIIGGGDGGVLREVVKHPSVESVVQCEIDEDVIQVSKKFLPGMAIGYSSSKLTLHVGDGFEFMKQNQDAFDVIITDSSDPMGPAESLFKESYYQLMKTALKEDGVLCCQGECQWLHLDLIKEMRQFCQSLFPVVAYAYCTIPTYPSGQIGFMLCSKNPSTNFQEPVQPLTQQQVAQMQLKYYNSDVHRAAFVLPEFARKALNDVS | |
RUO | |
SRM | |
Wheat Germ (in vitro) | |
GST | |
Liquid |